5-Chloro-2-(methylamino)benzophenone 99%
Catalog # L06338.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
CAS Number:1022-13-5
:
MDL Number:MFCD00008284
:245.71 g/mol
Melting Pt:93…98 °C
Storage Temperature:Ambient
EINECS:213-822-1
Formula:C₁₄H₁₂ClNO