
Phenacyltriphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAP1182-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:461.3301 g/mol
EINECS:227-945-3
:GHS07
CAS Number:6048-29-9
Formula:C₂₆H₂₂BrOP
MDL Number:MFCD00011920