Order Entry
Netherlands
ContactUsLinkComponent
2,4-Di-tert-butoxy-5-pyrimidineboronic acid hydrate
2,4-Di-tert-butoxy-5-pyrimidineboronic acid hydrate
Catalog # MAYBSB02090.250
Supplier:  Thermo Scientific
CAS Number:  
2,4-Di-tert-butoxy-5-pyrimidineboronic acid hydrate
Catalog # MAYBSB02090.250
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
CAS Number:306935-93-3
Formula:C₁₂H₂₁BN₂O₄·H₂O
MDL Number:MFCD02180675