
(Cyanomethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
: TCI
:4336-70-3
:
:C₂₀H₁₇ClNP
:MFCD00031672
:337.79 g/mol
:224-383-0
:3439
:6.1,III
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Frequently Bought Together