

2-(Trimethylsilyl)ethoxymethyl chloride ≥95.0% (by GC) stabilised
: TCI
:76513-69-4
:
:C₆H₁₅ClOSi
:MFCD00009919
:166.72 g/mol
:59°C
:46°C
:278-483-4
:2920
:8,II
:Stabilised with diisopropylethylamine
:GHS02
:GHS05
:Refrigerator
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together