

2-(Trimethylsilyl)ethoxymethyl chloride 90% tech. stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76513-69-4
:
Formula:C₆H₁₅ClOSi
MDL Number:MFCD00009919
:166.72 g/mol
Boiling Pt:57…59 °C (8 mmHg)
Density:0.94 g/cm³
Flash Pt:46 °C
Storage Temperature:Refrigerator
EINECS:278-483-4
UN:2920
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4380 (20°C, 589 nm)
- GC:≥88 %
- Stabilizer:≤200 ppm Diisopropylethylamine
Specification Test Results
Frequently Bought Together