
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:132705-51-2
:
Formula:C₁₂H₂₄BrF₆N₃P₂
MDL Number:MFCD00077412
:466.2 g/mol
:GHS07
Related Information
Frequently Bought Together