Order Entry
Netherlands
ContactUsLinkComponent
Cinnamyltriphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Irritant
Cinnamyltriphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Catalog # TCIAC1286-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Cinnamyltriphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Catalog # TCIAC1286-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Argentmetric Titration):
    min. 96.0 %
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :459.3600 g/mol
 :GHS07
CAS Number:7310-74-9
Formula:C₂₇H₂₄BrP
MDL Number:MFCD00040767