N(α)-Cbz-D-histidine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:67424-93-5
:
Formula:C₁₄H₁₅N₃O₄
MDL Number:MFCD00067702
:289.3 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+22.0 to +26.0 deg (C=6, 6mol/L HCl)
Specification Test Results
Frequently Bought Together