Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Chlorfenapyr 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Chlorfenapyr 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL11247500CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Chlorfenapyr 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL11247500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    122453-73-0
  • Description:
    Chlorfenapyr 10 µg/ml in cyclohexane
  • Formula:
    C₁₅H₁₁BrClF₃N₂O
  • Description:
    Chlorfenapyr

Specifications

About this item