Organic reference standard, Perylene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20915000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:198-55-0
- Description:Perylene 10 µg/ml in cyclohexane
- Formula:C₂₀H₁₂
- Description:Perylene