Bestelling invoeren
Netherlands
ContactUsLinkComponent
D-(+)-Glucurono-6,3-lactone
D-(+)-Glucurono-6,3-lactone
Catalog # ROTH9946.1
Supplier:  Roth Carl
CAS Number:  
D-(+)-Glucurono-6,3-lactone
Catalog # ROTH9946.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :176.12 g/mol
EINECS:251-053-3
CAS Number:32449-92-6
Formula:C₆H₈O₆
MDL Number:MFCD00135622