Order Entry
Netherlands
ContactUsLinkComponent
Borane trimethylamine complex ≥97.0% (by titrimetric analysis)
Flammable
Irritant
Borane trimethylamine complex ≥97.0% (by titrimetric analysis)
Catalog # TCIAT1181-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Borane trimethylamine complex ≥97.0% (by titrimetric analysis)
Catalog # TCIAT1181-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Melting point:
    93.0 to 97.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :72.95 g/mol
EINECS:200-850-4
UN:1325
ADR:4.1,II
 :GHS02
 :GHS07
CAS Number:75-22-9
Formula:C₃H₁₂BN
MDL Number:MFCD00012420