Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Borane trimethylamine complex 97%

Flammable
Irritant
Borane trimethylamine complex 97%
Certificates
Borane trimethylamine complex 97%
Borane trimethylamine complex 97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 75-22-9 Molecular Formula: (CH3)3N.BH3 Formula Weight: 72.95 Flash Point: 64°(147°F) Melting Point: 93-95° Boiling Point: 172° Beilstein Reference: 3905060 UN No.: 1325 MDL No.: MFCD00012420

  • CAS Number:75-22-9
  • Formula:C₃H₁₂BN
  • MDL Number:MFCD00012420
  • Molecular Weight:72.95 g/mol
  • Boiling Pt:172 °C
  • Melting Pt:93…95 °C
  • Density:0.81
  • Flash Pt:64 °C (147 °F)
  • Storage Temperature:Ambient
  • EINECS:200-850-4
  • UN:1325
  • ADR:4.1,II
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.