Borane trimethylamine complex 97%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:75-22-9
- Formula:C₃H₁₂BN
- MDL Number:MFCD00012420
- Molecular Weight:72.95 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
- EINECS:200-850-4
- UN:1325
- ADR:4.1,II
2 Options Available Below
Product Details & Documents
- CAS Number:75-22-9
- Formula:C₃H₁₂BN
- MDL Number:MFCD00012420
- Molecular Weight:72.95 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
- EINECS:200-850-4
- UN:1325
- ADR:4.1,II
Documents
ACRO40274
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:92 to 96 °C
- Titration with Na2S2O3:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



