
3,6,9-Trioxa-1,11-undecanediamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAD3664-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:192.2560 g/mol
Boiling Pt:127°C
EINECS:213-206-2
:GHS05
CAS Number:929-75-9
Formula:C₈H₂₀N₂O₃
MDL Number:MFCD00628164