
1,3-Dicyclohexyl-2-thiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1212-29-9
:MFCD00792562
Formula:C₁₃H₂₄N₂S
MDL Number:MFCD00021316
:240.41 g/mol
EINECS:214-920-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:181.0 to 184.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together