Glycine hydrochloride (H-Gly-OH.HCl) ≥99.0% (by titrimetric analysis)
Catalog # TCIAG0103-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Purity (Argentmetric Titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:111.53 g/mol
EINECS:227-841-8
CAS Number:6000-43-7
Formula:C₂H₆ClNO₂
MDL Number:MFCD00012872