Order Entry
Netherlands
ContactUsLinkComponent
2-Ethylhexyl-4-(dimethylamino)benzoate
2-Ethylhexyl-4-(dimethylamino)benzoate
Catalog # EHERC15841000
Supplier:  EHRENSTORFER
CAS Number:  
2-Ethylhexyl-4-(dimethylamino)benzoate
Catalog # EHERC15841000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :277.40 g/mol
EINECS:244-289-3
CAS Number:21245-02-3
Formula:C₁₇H₂₇NO₂
MDL Number:MFCD00017526