Herbicide reference standard, Propisochlor 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL16495000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:86763-47-5
- Description:Propisochlor 10 µg/ml in cyclohexane
- Formula:C₁₅H₂₂ClNO₂
- Description:Propisochlor