
4,6-Dimethoxysalicylaldehyde ≥98.0% (by GC, titration analysis)
: TCI
:708-76-9
:
:C₉H₁₀O₄
:MFCD00003328
:182.1733 g/mol
:131°C
:211-904-1
:GHS07
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
Frequently Bought Together