Culture flasks, DeLong®
These DeLong® culture flasks are made of borosilicate glass type 1, class A.
- Ideal for growing, storing and mixing of aerobic and anaerobic cultures
- Graduated to approximate volumes
- Very good chemical and temperature resistance
: The stainless steel or KAP-UTS™ disposable closures must be purchased separately for the flasks without closures.
Stainless steel caps without fingers
Frequently Bought Together