Eppendorf® Easypet® 3 Electronic Pipette Controller
Catalog # 613-2795
Supplier: EPPENDORF
Specifications
- Description:Easypet® 3
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Pk:1
Specifications
About this item
Electronic pipetting doesn't come any more advanced than the Easypet 3 from Eppendorf. Designed to work with Eppendorf serological pipettes, you get complete speed control and utmost precision in an extremely light and well-balanced body that fits comfortably in your hand, for fatigue-free pipetting.
- Intuitive and convenient speed adjustment simply done with the tips of your fingers
- Lightweight, well-balanced and ergonomic design that allows for fatigue-free pipetting
- Vibrant backlit LEDs provide optical feedback of the remaining battery life
- Lithium polymer rechargeable battery offers long cordless runtime
- Smooth setting of pump speed
- Operation while recharging is possible
- Autoclavable pipette adapter for sterile applications
- Quick release of aspirating cone for easy exchange of membrane filters
The lightweight, well-balanced Easypet 3 has been designed considering all ergonomic findings to fit comfortably in your hand and matches its shape to allow for fatigue-free pipetting.
Electronic pipette controller with air cushion system with autoclavable pipette adapter and aspirating cone.
Ordering information: The Easypet® 3 Pipette Controller is supplied with mains/power supply, wall mounting device, shelf stand, and two 0.45 µm membrane filters.