Order Entry
Netherlands
ContactUsLinkComponent
VWR®, Digital Timer, One Channel
VWR®, Digital Timer, One Channel
Catalog # 609-0185
Supplier:  VWR Collection
undefined
VWR®, Digital Timer, One Channel
Catalog # 609-0185
Supplier:  VWR Collection

Specifications

  • Colour:
    White
  • Description:
    Digital timer, single channel
  • W×D×H:
    65×15×67 mm
  • Accuracy:
    ±0,01% (1 s)
  • Pk:
    1

Specifications

About this item

Counts down from 99 minutes, 59 seconds.

  • Loud alarm for noisy environments
  • Memory function
  • Small, portable unit
  • Automatic off function
Delivery information: Supplied with magnet, stand, clip and certificate of calibration.