Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,7-Diaza-15-crown-5 (1,4,10-trioxa-7,13-diazacyclopentadecane, Kryptofix® 21) ≥97.0% (by GC)
  TCIAD2744-1G
 :  TCI
 :  
undefined
 
1,7-Diaza-15-crown-5 (1,4,10-trioxa-7,13-diazacyclopentadecane, Kryptofix® 21) ≥97.0% (by GC)
  TCIAD2744-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    85.0 to 92.0 deg-C

 

 

 :
 :218.3 g/mol
 :250-530-3
 :GHS07
 :31249-95-3
 :C₁₀H₂₂N₂O₃
 :MFCD00005109