



Benzoyl peroxide blend with 50% dicyclohexyl phthalate, Sigma-Aldrich®
Catalog # B5907-1VL
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Appearance (Colour):White (May contain off-coloured particles)
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
Specification Test Results
About this item
:
:242.23 g/mol
Melting Pt:54 °C
EINECS:202-327-6
ADR:5.2,
:M-Clarity™ quality level = MQ200
: 50%
UN:3104
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
CAS Number:94-36-0
Formula:C₁₄H₁₀O₄
MDL Number:MFCD00003071