Order Entry
Netherlands
ContactUsLinkComponent
Wright's stain for microscopy and bacteriology
Wright's stain for microscopy and bacteriology
Supplier:  TCI
Certificates
Wright's stain for microscopy and bacteriology
Supplier:  TCI

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Lambda max.:
    521.0 to 526.0 nm (MeOH)
  • Lambda max.2:
    637.0 to 643.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 700 (MeOH, 521.0 to 526.0 nm)
  • Absorbance (E1%1cm)2:
    min. 1100 (MeOH, 637.0 to 643.0 nm)

Specification Test Results

Related Information

Frequently Bought Together