Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dibutyl-2-thiourea ≥97.0% (by total nitrogen basis)
Irritant
1,3-Dibutyl-2-thiourea ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Irritant
1,3-Dibutyl-2-thiourea ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    64.0 to 68.0 deg-C
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

Frequently Bought Together