L(-)-Malic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-67-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00064213
:134.09 g/mol
Merck Index:14,5707
Boiling Pt:322 °C (1013 hPa)
Melting Pt:100…106 °C
Density:1.6
Flash Pt:220 °C (428 °F)
EINECS:202-601-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together