Order Entry
Netherlands
ContactUsLinkComponent
Lactic acid
Lactic acid
Catalog # EHERC14582000
Supplier:  EHRENSTORFER
CAS Number:  
Lactic acid
Catalog # EHERC14582000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Merck Index:13,05351
 :90.08 g/mol
EINECS:200-018-0
 :GHS05
Storage Temperature:Ambient
CAS Number:50-21-5
 :598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520