
1-Octanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:5324-84-5
:
Formula:C₈H₁₇NaO₃S
MDL Number:MFCD00007544
:216.28 g/mol
Melting Pt:> 300 °C
EINECS:226-195-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Absorbance (210nm):max. 1.00
- Absorbance (230nm):max. 0.30
- Absorbance (254nm):max. 0.13
Specification Test Results
Frequently Bought Together