
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.9% for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Merck Index:14,9772
Boiling Pt:219…220 °C (10 mmHg)
Melting Pt:168…172 °C
Density:1.353
EINECS:201-064-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together