Order Entry
Netherlands
ContactUsLinkComponent
 
Corrosive
Ethyl-6-(chloroformyl)hexanoate ≥97.0% (by titrimetric analysis)
Catalog # TCIAC1414-5G
Supplier:  TCI
CAS Number:  
Corrosive
Ethyl-6-(chloroformyl)hexanoate ≥97.0% (by titrimetric analysis)
Catalog # TCIAC1414-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (Argentmetric Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
Boiling Pt:141°C
UN:3265
ADR:8,III
 :206.67 g/mol
 :GHS05
CAS Number:14794-32-2
Formula:C₉H₁₅ClO₃
MDL Number:MFCD00671507