
cis-1,4-Cyclohexanedicarboxylic acid ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:619-81-8
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00064950
:172.1800 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 90.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:167.0 to 172.0 deg-C
Specification Test Results
Frequently Bought Together