Sodium hexanoate ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10051-44-2
:
Formula:C₆H₁₁NaO₂
MDL Number:MFCD00059056
:138.1401 g/mol
EINECS:233-179-0
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Nonaqueous Titration):99.0 to 101.0 % (calcd.on dried substance)
- Solubility in Water:almost transparency
- Drying loss:max. 3.0 %
Specification Test Results
Frequently Bought Together