Digitonin, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:11024-24-1
:
Formula:C₅₆H₉₂O₂₉
MDL Number:MFCD00077729
:1229.312 g/mol
EINECS:234-255-6
UN:3462
ADR:6.1,III
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Serological pipettes, Stripette™
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection