Order Entry
Netherlands
ContactUsLinkComponent
2,2-Dimethoxy-2-phenylacetophenone ≥98.0% (by GC)
Irritant
Environment
2,2-Dimethoxy-2-phenylacetophenone ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Environment
2,2-Dimethoxy-2-phenylacetophenone ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    65.0 to 69.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together