
4-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:182344-23-6
:
Formula:C₇H₅BF₄O₂
MDL Number:MFCD07363787
:207.92 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together