Order Entry
Netherlands
ContactUsLinkComponent
3-Fluoro-4-methylphenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Irritant
3-Fluoro-4-methylphenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
Irritant
3-Fluoro-4-methylphenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 113.0 %

Specification Test Results

Related Information

Frequently Bought Together