Order Entry
Netherlands
ContactUsLinkComponent
Sodium-1-hexadecanesulphonate for synthesis, Sigma-Aldrich®
Sodium-1-hexadecanesulphonate for synthesis, Sigma-Aldrich®
Catalog # 8.41813.0010
Supplier:  Merck
CAS Number:  
Sodium-1-hexadecanesulphonate for synthesis, Sigma-Aldrich®
Catalog # 8.41813.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Melting Point:
    265.3 °C
  • Molar Mass:
    328.48 g/mol
  • Hill Formula:
    C16H33NaO3S

Specification Test Results

About this item

 :
 :328.49 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:239-100-6
CAS Number:15015-81-3
Formula:C₁₆H₃₃NaSO₃
MDL Number:MFCD00007528