3-Pyridinesulphonic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:636-73-7
:
Formula:C₅H₅NO₃S
MDL Number:MFCD00006381
:159.16 g/mol
Melting Pt:360°C
EINECS:211-265-9
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:almost transparency
- Water:max. 1.0 %
Specification Test Results
Frequently Bought Together