3-Pyridinesulphonic acid
Catalogus # MOLEM30531969
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:159.16 g/mol
EINECS:211-265-9
CAS Number:636-73-7
Formula:C₅H₅NO₃S
MDL Number:MFCD00006381