

2-Bromo-4'-(trifluoromethyl)acetophenone ≥95.0% (by GC)
: TCI
:383-53-9
:
:C₉H₆BrF₃O
:MFCD00126489
:267.0400 g/mol
:3261
:8,III
:GHS05
:GHS07
:Refrigerator
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:53.0 to 57.0 deg-C
Frequently Bought Together