
Yttrium(III) 2-ethylhexanoate ≥99.8% (metals basis)
Catalog # 36661.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
CAS Number:103470-68-4
:
Formula:C₂₄H₄₅O₆Y
MDL Number:MFCD00070441
:518.524 g/mol
:GHS08
Storage Temperature:Ambient