Order Entry
Netherlands
ContactUsLinkComponent
2-Amino-5,6-dimethylbenzothiazole ≥98.0% (by HPLC, titration analysis)
Irritant
2-Amino-5,6-dimethylbenzothiazole ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
2-Amino-5,6-dimethylbenzothiazole ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    187.0 to 192.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together