

1,4-Thioxane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:15980-15-1
:
Formula:C₄H₈OS
MDL Number:MFCD00006573
:104.17 g/mol
Boiling Pt:67°C
Flash Pt:42°C
EINECS:240-117-6
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together