
Tetrahydropyran-2-methanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:100-72-1
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00006624
:116.16 g/mol
Boiling Pt:187°C
Flash Pt:93°C
EINECS:202-882-4
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together