

1,4-Dioxene ≥98.0% (by GC)
: TCI
:543-75-9
:
:C₄H₆O₂
:MFCD00082537
:86.09 g/mol
:94°C
:1°C
:1993
:3,II
:GHS02
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together