
3-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:175883-63-3
:
Formula:C₇H₈BClO₂
MDL Number:MFCD04039010
:170.40 g/mol
Melting Pt:247°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
Related Information
Frequently Bought Together