Herbicide reference standard, Acetochlor 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA10018000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:34256-82-1
- Description:Acetochlor 100 µg/ml in cyclohexane
- Formula:C₁₄H₂₀ClNO₂
- Description:Acetochlor